missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-omega-Monomethyl-L-arginine monoacetate, 99%

CAS: 53308-83-1 | C9H20N4O4 | 248.283 g/mol

$552.13 - $552.13

Chemical Identifiers

CAS 53308-83-1
Molecular Formula C9H20N4O4
Molecular Weight (g/mol) 248.283
MDL Number MFCD00069311
InChI Key IKPNWIGTWUZCKM-JEDNCBNOSA-N
Synonym tilarginine acetate, l-nmma, l-nmma acetate, ng-monomethyl-l-arginine acetate, nomega-me-l-arg, unii-2fl3530af2, ng-me-l-arg, acoh, tilarginine acetate usan, ng-methyl-l-arginine acetate salt, l-ornithine, n5-imino methylamino methyl-cas
PubChem CID 135242
IUPAC Name acetic acid;(2S)-2-amino-5-[(N'-methylcarbamimidoyl)amino]pentanoic acid
SMILES CC(=O)O.CN=C(N)NCCCC(C(=O)O)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC328721000
SDS View Documents
Thermo Scientific Chemicals
328721000
100 mg Glass Bottle
Each for $552.13
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

53308-83-1
248.283
IKPNWIGTWUZCKM-JEDNCBNOSA-N
135242
CC(=O)O.CN=C(N)NCCCC(C(=O)O)N
C9H20N4O4
MFCD00069311
tilarginine acetate, l-nmma, l-nmma acetate, ng-monomethyl-l-arginine acetate, nomega-me-l-arg, unii-2fl3530af2, ng-me-l-arg, acoh, tilarginine acetate usan, ng-methyl-l-arginine acetate salt, l-ornithine, n5-imino methylamino methyl-cas
acetic acid;(2S)-2-amino-5-[(N'-methylcarbamimidoyl)amino]pentanoic acid

Specifications

180°C to 190°C
98.5
White
99%
Glass Bottle
+9° to +13° (20°C, 589nm) (c=4, water)
tilarginine acetate, l-nmma, l-nmma acetate, ng-monomethyl-l-arginine acetate, nomega-me-l-arg, unii-2fl3530af2, ng-me-l-arg, acoh, tilarginine acetate usan, ng-methyl-l-arginine acetate salt, l-ornithine, n5-imino methylamino methyl-cas
CC(=O)O.CN=C(N)NCCCC(C(=O)O)N
+ 10.50 (20.00°C c=4,water)
248.283
248.3
Powder
53308-83-1
100.0
Authentic
C9H20N4O4
MFCD00069311
15, 6096
IKPNWIGTWUZCKM-JEDNCBNOSA-N
acetic acid;(2S)-2-amino-5-[(N'-methylcarbamimidoyl)amino]pentanoic acid
100 mg
135242
≥98.5%
N-ω-Monomethyl-L-arginine monoacetate
Videos
SDS
Documents

Documents

RUO – Research Use Only