accessibility menu, dialog, popup

UserName

H3CNNH2HN(S)NH2OHOCH3OHO(E/Z)

CAS RN 53308-83-1

IUPAC Name: (2S)-2-amino-5-(N''-methylcarbamimidamido)pentanoic acid; acetic acid
Synonyms: (2S)-2-amino-5-(N''-methylcarbamimidamido)pentanoic acid; acetic acid
Molecular Weight (g/mol): 248.28
Molecular Formula: C9H20N4O4
InChi Key: IKPNWIGTWUZCKM-JEDNCBNOSA-N
SMILES: CC(O)=O.CN=C(N)NCCC[C@H](N)C(O)=O
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)