missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-omega-Monomethyl-L-arginine monoacetate, 99%

Catalog No. AC328721000
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC328721000 100 mg Glass Bottle
AC328720050 5 mg Glass Bottle
AC328720250 25 mg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC328721000 Supplier Thermo Scientific Chemicals Supplier No. 328721000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 53308-83-1 | C9H20N4O4 | 248.283 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 53308-83-1
Molecular Formula C9H20N4O4
Molecular Weight (g/mol) 248.283
MDL Number MFCD00069311
InChI Key IKPNWIGTWUZCKM-JEDNCBNOSA-N
Synonym tilarginine acetate, l-nmma, l-nmma acetate, ng-monomethyl-l-arginine acetate, nomega-me-l-arg, unii-2fl3530af2, ng-me-l-arg, acoh, tilarginine acetate usan, ng-methyl-l-arginine acetate salt, l-ornithine, n5-imino methylamino methyl-cas
PubChem CID 135242
IUPAC Name acetic acid;(2S)-2-amino-5-[(N'-methylcarbamimidoyl)amino]pentanoic acid
SMILES CC(=O)O.CN=C(N)NCCCC(C(=O)O)N

Specifications

Melting Point 180°C to 190°C
CAS Min % 98.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Specific Rotation +9° to +13° (20°C, 589nm) (c=4, water)
Merck Index 15, 6096
Specific Rotation Condition + 10.50 (20.00°C c=4,water)
Quantity 100 mg
Formula Weight 248.3
Percent Purity ≥98.5%
Physical Form Powder
Chemical Name or Material N-ω-Monomethyl-L-arginine monoacetate
Show More Show Less

RUO – Research Use Only