Learn More
Nextteq™ Gastec™ Dosimeter Tubes
Catalog No. 19026357
Change view
Click to view available options
Components Measured:
Acetaldehyde, Formaldehyde, Furfural, Methyl Ethyl Ketone
Catalog No. Components Measured
19026357 Acetaldehyde, Formaldehyde, Furfural, Methyl Ethyl Ketone
1 options
Catalog No. 19026357 Supplier Nextteq™ Supplier No. 91D
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Components Measured Acetaldehyde, Formaldehyde, Furfural, Methyl Ethyl Ketone
Measurement Range Acetaldehyde: 0.1-20ppm, Formaldehyde: 0.10-20ppm, Furfural: 0.3-60ppm, Methyl Ethyl Ketone: 0.125-25ppm
Quantity 10
Response Time Acetaldehyde: 1-10 hr., Formaldehyde: 1-10 hr., Furfural: 1-10 hr., Methyl Ethyl Ketone: 1-10 hr.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C