missing translation for 'onlineSavingsMsg'
Learn More

Nextteq™ Gastec™ Detector Tubes: Water Vapor

Each tube contains a reagent that reacts with a specific gas/vapor, yielding a stain that is measured against calibrations printed on the tube.

$346.98 - $566.60

Specifications

Components Measured Water Vapor
Quantity 10
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C
View More Specs

Products 2
Catalog Number Mfr. No. Measurement Range Response Time No. of Strokes Price Quantity  
Catalog Number Mfr. No. Measurement Range Response Time No. of Strokes Price Quantity  
19-026-321
View Documents
Nextteq™
6L
0.05 to 2mg/L 15 to 30 sec. 0.5 to 1 N/A
Only null left
 
19-026-322
View Documents
Nextteq™
6LLP
2 to 10LB/MMCF 3 min. 2 N/A
Only null left
 
Description

Description

Each tube contains a reagent that reacts with a specific gas/vapor, yielding a stain that is measured against calibrations printed on the tube.

Gastec tubes offer accurate, effective readings of more than 500 gases and vapors. Each glass detector tube contains a reagent which reacts with a specific gas or vapor, yielding a stain that is measured against calibrations printed on the tube. The unique absorbing media produce extremely clear lines of stain demarcation for precise reading, and are stable, with a shelf-life of up to two years, depending on the application. (See below for tube selection of specific gases and vapors.) Sold in packs of 10.
Specifications

Specifications

Water Vapor
10
32°F to 104°F
0°C to 40°C
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'