missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nextteq™ Gastec™ Detector Tubes: Formaldehyde
Each tube contains a reagent that reacts with a specific gas/vapor, yielding a stain that is measured against calibrations printed on the tube.
$562.18
Specifications
| Quantity | 10 |
|---|---|
| Temperature Range (English) | 32°F to 104°F |
| Temperature Range (Metric) | 0°C to 40°C |
| Catalog Number | Mfr. No. | Components Measured | Measurement Range | Response Time | No. of Strokes | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Components Measured | Measurement Range | Response Time | No. of Strokes | Price | Quantity & Availability | ||
|
19026358
|
Nextteq™
91L |
Formaldehyde | 0.1 to 40 | 1.5 to 7.5 min. | 1 to 5 |
Pack of 10 Each for $562.18
Case of 10 Pack for $5,621.80
|
|
||
Description
Each tube contains a reagent that reacts with a specific gas/vapor, yielding a stain that is measured against calibrations printed on the tube.
Gastec tubes offer accurate, effective readings of more than 500 gases and vapors. Each glass detector tube contains a reagent which reacts with a specific gas or vapor, yielding a stain that is measured against calibrations printed on the tube. The unique absorbing media produce extremely clear lines of stain demarcation for precise reading, and are stable, with a shelf-life of up to two years, depending on the application. (See below for tube selection of specific gases and vapors.) Sold in packs of 10.Specifications
| 10 | |
| 32°F to 104°F | |
| 0°C to 40°C |