missing translation for 'onlineSavingsMsg'
Learn More

Nextteq™ Gastec™ Detector Tubes: Ammonia

Each tube contains a reagent that reacts with a specific gas/vapor, yielding a stain that is measured against calibrations printed on the tube.

$193.84

Specifications

Components Measured Ammonia
Quantity 10
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C
View More Specs

Products 2
Catalog Number Mfr. No. Measurement Range Response Time No. of Strokes Price Quantity & Availability  
Catalog Number Mfr. No. Measurement Range Response Time No. of Strokes Price Quantity & Availability  
19026281
View Documents Promotion Details
Nextteq™
3M
10 to 1000 20 sec. to 1 min. 0.5 to 5
Each for $193.84
Only null left
 
19026279
View Documents Promotion Details
Nextteq™
3L
0.5 to 78 30 sec. to 1.5 min. 0.5 to 2
Pack of 10 Each for $193.84
Case of 10 Pack for $5,621.80
Only null left
 
Description

Description

Each tube contains a reagent that reacts with a specific gas/vapor, yielding a stain that is measured against calibrations printed on the tube.

Gastec tubes offer accurate, effective readings of more than 500 gases and vapors. Each glass detector tube contains a reagent which reacts with a specific gas or vapor, yielding a stain that is measured against calibrations printed on the tube. The unique absorbing media produce extremely clear lines of stain demarcation for precise reading, and are stable, with a shelf-life of up to two years, depending on the application. (See below for tube selection of specific gases and vapors.) Sold in packs of 10.
Specifications

Specifications

Ammonia
10
32°F to 104°F
0°C to 40°C
Videos
SDS
Documents

Documents