Learn More
Sartorius NexaGel Cell Recovery Solution
Catalog No. 14555859
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14555859 500 mL
14555858 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14555859 Supplier Sartorius Supplier No. NGR04500
Only null left
Non-enzymatic, ready-to-use cell recovery solution designed for the rapid and efficient harvesting of organoids or 3D cells cultured with NexaGel™or animal-based ECM.
  • Stable solution at room temperature with neutral pH, and also maintains cell integrity
  • Recovered cells can be easily sub-cultured in both 2D and 3D formats
  • The solution is versatile and can be used before or after the fixation and staining of hydrogel specimens, ensuring high-quality data for downstream analysis
  • It is suitable for harvesting cells from 2D ECM-coated plates
  • Ensures a high recovery rate and cell viability, making it perfect for passaging, cryopreservation, or further biochemical analysis
Specifications
Form Liquid
Product Type Organoid Harvesting Solution
Sterility Sterile
Sterilization Method Autoclaved
Quantity 500 mL