Learn More
Sartorius NexaGel Cell Recovery Solution
Description
- Stable solution at room temperature with neutral pH, and also maintains cell integrity
- Recovered cells can be easily sub-cultured in both 2D and 3D formats
- The solution is versatile and can be used before or after the fixation and staining of hydrogel specimens, ensuring high-quality data for downstream analysis
- It is suitable for harvesting cells from 2D ECM-coated plates
- Ensures a high recovery rate and cell viability, making it perfect for passaging, cryopreservation, or further biochemical analysis
Specifications
Specifications
| Form | Liquid |
| Product Type | Organoid Harvesting Solution |
| Sterility | Sterile |
| Sterilization Method | Autoclaved |
| Quantity | 100 mL |