missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rubbermaid™ Indoor Station Containers and Tops
$1,054.39 - $1,457.25
| Catalog Number | Mfr. No. | Capacity (English) | Dimensions (L x W x H) | Material | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (English) | Dimensions (L x W x H) | Material | Price | Quantity | |||
|
19-813-111
|
Rubbermaid™
395873BLUE |
35 gal. | 19.5 x 19.5 x 27.6 in. | Plastic |
N/A
|
N/A | |||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
19-813-120
|
Rubbermaid™
396873DBLUE |
50 gal. | 24.75 x 25.2 x 42.1 in. | Polyethylene |
N/A
|
N/A | |||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
19-813-121
|
Rubbermaid™
396973DBLUE |
50 gal. | 24.75 x 25.2 x 42.1 in. | Resin |
N/A
|
N/A | |||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Specifications
| Blue |