missing translation for 'onlineSavingsMsg'
Learn More

New Pig Grippy™ Absorbent Mat Roll

Catalog No. 19039255
Change view
Click to view available options
Dimensions (L x W):
50 ft. x 32 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19039255 50 ft. x 32 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19039255 Supplier New Pig Supplier No. MAT3250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mat absorbs leaks, keeping work spaces safe

 

  • Spunbond top layer with zigzag stitching stands up to foot and light-wheeled traffic, as well as heavy equipment drive-overs
  • Eight layers of fine fiber polypropylene safely absorb leaks, drips and overspray of oil, coolants, solvents and water
  • Full-coverage adhesive backing holds tight but leaves no residue
  • Reduces trip hazards by lying flat without curling or bunching
  • Pells up easily for quick change-outs
  • Dark color hides grime
  • 81.3cm × 15.24m (32 × 50 ft)

 

Certifications

Tested and certified by National Floor Safety

Specifications

Color Blue
Material Absorbent: Polypropylene; Stitching: Polyester; Adhesive Backing: Proprietary Adhesive Material
Type Adhesive-backed Absorbent Mat
Dimensions (L x W) 50 ft. x 32 in.
Length (English) 50 ft.
Length (Metric) 15.24 m
Width (English) 32 in.
Width (Metric) 81.28 cm