missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals New methylene Blue N, pure

Catalog No. AC415530025
Change view
Click to view available options
Quantity:
2.5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC415530025 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC415530025 Supplier Thermo Scientific Chemicals Supplier No. 415530025
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1934-16-3 | C18H22ClN3S | 347.905 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1934-16-3
Molecular Formula C18H22ClN3S
Molecular Weight (g/mol) 347.905
MDL Number MFCD00135878
InChI Key SQHOAFZGYFNDQX-UHFFFAOYSA-N
Synonym Basic Blue 24
PubChem CID 73518
IUPAC Name ethyl-[7-(ethylamino)-2,8-dimethylphenothiazin-3-ylidene]azanium;chloride
SMILES CCNC1=CC2=C(C=C1C)N=C3C=C(C(=[NH+]CC)C=C3S2)C.[Cl-]

Specifications

Dye Content ca 70%
Infrared Spectrum Authentic
Packaging Glass bottle
Beilstein 27, IV, 5163
Color Brown to Green
Quantity 2.5 g
Physical Form Crystalline Powder or Crystals
Formula Weight 347.9
Grade Pure
Chemical Name or Material New methylene Blue N, 85%

RUO – Research Use Only