missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals New Coccine, pure
CAS: 2611-82-7 | C20H11N2Na3O10S3 | 604.46 g/mol
$71.50 - $223.45
Chemical Identifiers
| CAS | 2611-82-7 |
|---|---|
| Molecular Formula | C20H11N2Na3O10S3 |
| Molecular Weight (g/mol) | 604.46 |
| MDL Number | MFCD00004084 |
| InChI Key | IVNZBWNBYXERPK-DZGBHZPSSA-K |
| Synonym | Acid Red 18, C.I. 16255, Cochineal Red A |
| PubChem CID | 131851859 |
| IUPAC Name | (8Z)-7-oxo-8-[(4-sulfonaphthalen-1-yl)hydrazinylidene]naphthalene-1,3-disulfonic acid;sodium |
| SMILES | [Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC2=C(\C(=N\NC3=C4C=CC=CC4=C(C=C3)S([O-])(=O)=O)C(=O)C=C2)C(=C1)S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC190230250
|
Thermo Scientific Chemicals
190230250 |
25 g | Glass bottle |
Each for $71.50
|
|
||||
|
AC190231000
|
Thermo Scientific Chemicals
190231000 |
100 g | Glass bottle |
Each for $223.45
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2611-82-7 | |
| 604.46 | |
| IVNZBWNBYXERPK-DZGBHZPSSA-K | |
| 131851859 | |
| [Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC2=C(\C(=N\NC3=C4C=CC=CC4=C(C=C3)S([O-])(=O)=O)C(=O)C=C2)C(=C1)S([O-])(=O)=O |
| C20H11N2Na3O10S3 | |
| MFCD00004084 | |
| Acid Red 18, C.I. 16255, Cochineal Red A | |
| (8Z)-7-oxo-8-[(4-sulfonaphthalen-1-yl)hydrazinylidene]naphthalene-1,3-disulfonic acid;sodium |
Specifications
| 2611-82-7 | |
| C20H11N2Na3O10S3 | |
| 16, I, 306 | |
| IVNZBWNBYXERPK-DZGBHZPSSA-K | |
| [Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC2=C(\C(=N\NC3=C4C=CC=CC4=C(C=C3)S([O-])(=O)=O)C(=O)C=C2)C(=C1)S([O-])(=O)=O | |
| 604.46 | |
| 10% max. (135°C, 6 hrs) | |
| Extra Pure | |
| Complies | |
| 25 g | |
| New Coccine |
| 75% min. | |
| MFCD00004084 | |
| Acid Red 18, C.I. 16255, Cochineal Red A | |
| Authentic | |
| (8Z)-7-oxo-8-[(4-sulfonaphthalen-1-yl)hydrazinylidene]naphthalene-1,3-disulfonic acid;sodium | |
| 131851859 | |
| 604.46 | |
| Glass bottle | |
| Bordeaux to Brown | |
| Powder |
Safety and Handling
EINECSNumber : 220-036-2
RTECSNumber : QJ6530000
TSCA : TSCA
RUO – Research Use Only