missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals New Coccine, pure

CAS: 2611-82-7 | C20H11N2Na3O10S3 | 604.46 g/mol

Supplier:  Thermo Scientific Chemicals 190230250

 View more versions of this product

Catalog No. AC190230250


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

New Coccine
75% min.
MFCD00004084
Acid Red 18, C.I. 16255, Cochineal Red A
Authentic
(8Z)-7-oxo-8-[(4-sulfonaphthalen-1-yl)hydrazinylidene]naphthalene-1,3-disulfonic acid;sodium
131851859
604.46
Glass bottle
Bordeaux to Brown
Powder
2611-82-7
C20H11N2Na3O10S3
16, I, 306
IVNZBWNBYXERPK-DZGBHZPSSA-K
[Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC2=C(\C(=N\NC3=C4C=CC=CC4=C(C=C3)S([O-])(=O)=O)C(=O)C=C2)C(=C1)S([O-])(=O)=O
604.46
10% max. (135°C, 6 hrs)
Extra Pure
Complies
25 g

Chemical Identifiers

2611-82-7
604.46
IVNZBWNBYXERPK-DZGBHZPSSA-K
131851859
[Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC2=C(\C(=N\NC3=C4C=CC=CC4=C(C=C3)S([O-])(=O)=O)C(=O)C=C2)C(=C1)S([O-])(=O)=O
C20H11N2Na3O10S3
MFCD00004084
Acid Red 18, C.I. 16255, Cochineal Red A
(8Z)-7-oxo-8-[(4-sulfonaphthalen-1-yl)hydrazinylidene]naphthalene-1,3-disulfonic acid;sodium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 220-036-2

RTECSNumber : QJ6530000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only