Learn More
Eppendorf™ Scientific Shaker Test Tube Platforms
For use with New Brunswick Innova™, I-Series™ and Excella™ Shakers
Supplier: Eppendorf™ M12890600
Description
Mount on universal platforms
Capacities listed are the most vessels of each size that will fit in a balanced pattern.
Chemical Identifiers
| Test Tube Rack | |
| Stainless Steel | |
| Innova 2100, 2150, 2300, 2350; Excella E10, E24/E24R, E25/E25R; all I-Series | |
| Large |
Specifications
| 26 to 30 mm | |
| Stainless Steel | |
| 20 |
| Test Tube Rack | |
| Innova 2100, 2150, 2300, 2350; Excella E10, E24/E24R, E25/E25R; all I-Series | |
| Large |