missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nevirapine Related Compound A Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Nevirapine Related Compound A is an impurity of nevirapine. Nevirapine is a non-nucleoside reverse transcriptase inhibitor (NNRTI), which belongs to the class of antiretroviral drugs. It acts by inhibiting the reverse transcriptase enzyme, thus stopping the multiplication of HIV. It is highly effective against advanced HIV disease.
Specifications
Specifications
| CAS | 133627-17-5 |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Linear Formula | C14H14N4 O |
| Synonym | 11-Ethyl-5,11-dihydro-4-methyl-6 H-dipyrido[3,2-b:2′,3′-e][1,4 ]diazepin-6-one; 5,11-DIHYDRO-6 H-11-ETHYL- 4-METHYL-DIPYRIDO[3,2-b:2′,3′- e][1,4 ]DIAZEPIN-6-ONE |
| Type | Certified Reference Material |
| Quantity | 50 mg |
| Formula Weight | 254.29 |
| Grade | Certified Reference Material |