missing translation for 'onlineSavingsMsg'
Learn More

Nevirapine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012915
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012915 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012915 Supplier Merck Emd Millipore Supplier No. PHR17571G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Nevirapine is a non-nucleoside reverse transcriptase inhibitor (NNRTI), which belongs to the class of antiretroviral drugs. It acts by inhibiting the reverse transcriptase enzyme, thus suppressing the multiplication of HIV. It is highly effective against the advanced HIV disease.

Specifications

CAS 129618-40-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C15H14N4 O
MDL Number MFCD00866928
Synonym Nevirapine; 11-Cyclopropyl-5,11-dihydro-4-methyl-6 H-dipyrido[3,2-b:2′,3′-e][1,4 ]diazepin-6-one
Type Certified Reference Material
Quantity 1 g
Formula Weight 266.30
Grade Certified Reference Material