missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NeutrAvidin Protein, DyLight™ 680

Catalog No. PI22848
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI22848 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI22848 Supplier Invitrogen™ Supplier No. 22848

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Protein

22848 contains 1 mL of protein in blue green colored liquid at a concentration of 1 mg/mL.

Avidin is a glycoprotein found in the egg white and tissues of birds, reptiles and amphibia. The biotin-binding protein contains four identical subunits having a combined mass of 67,000-68,000 daltons. Removing the glycosylation from avidin yields NeutrAvidin Protein with a mass of 60,000 daltons. Carbohydrate-based lectin-binding is reduced to undetectable levels, yet biotin-binding affinity is retained. NeutrAvidin Protein offers the advantages of a neutral pI to minimize nonspecific adsorption, along with lysine residues that remain available for derivatization or other customized conjugation. NeutrAvidin Protein yields the lowest nonspecific binding among the known biotin binding proteins. The specific activity for biotin-binding is approximately 14 μg/mg of protein, which is near the theoretical maximum activity.

Specifications

Concentration 1 mg/mL
For Use With (Application) ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Formulation PBS with 1% BSA and 0.02% sodium azide; pH 7.2
Name NeutrAvidin
Conjugate DyLight 680
pH Range 7.2
Cross Reactivity Tag
Purification Method Purified
Quantity 1 mL
Storage Requirements 4°C
Regulatory Status RUO
pH 7.2
Common Name NeutrAvidin
Product Type Protein
Form Liquid
Purity or Quality Grade purified
Host Species N/A
Show More Show Less