missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NeutrAvidin Protein, DyLight™ 550

Catalog No. 84606
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
84606 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 84606 Supplier Invitrogen™ Supplier No. 84606

Protein

NeutrAvidin for ICC/IF

Avidin is a glycoprotein found in the egg white and tissues of birds, reptiles and amphibia. The biotin-binding protein contains four identical subunits having a combined mass of 67,000-68,000 daltons. Removing the glycosylation from avidin yields NeutrAvidin Protein with a mass of 60,000 daltons. Carbohydrate-based lectin-binding is reduced to undetectable levels, yet biotin-binding affinity is retained. NeutrAvidin Protein offers the advantages of a neutral pI to minimize nonspecific adsorption, along with lysine residues that remain available for derivatization or other customized conjugation. NeutrAvidin Protein yields the lowest nonspecific binding among the known biotin binding proteins. The specific activity for biotin-binding is approximately 14 μg/mg of protein, which is near the theoretical maximum activity.

Specifications

Concentration 1mg/mL
Conjugate DyLight 550
Cross Reactivity Tag
Formulation Buffered saline solution with 1% BSA and no preservative
Quantity 1 mL
Regulatory Status RUO
Content And Storage Stable for 1 year at 4°C
Form Liquid
For Use With (Application) Immunofluorescence
Purity or Quality Grade purified
Host Species N/A
Product Type NeutrAvidin
Show More Show Less