missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NeutrAvidin Protein

Catalog No. PI31055
Change view
Click to view available options
Quantity:
10 mg
100 mg
1000 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI31055 1000 mg
PI31000 10 mg
PI31050 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PI31055 Supplier Thermo Scientific™ Supplier No. 31055

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

NeutrAvidin Protein, from Invitrogen. Species Reactivity: ; Applications: ELISA, Immunohistochemistry, In Situ Hybridization (ISH), Western Blot.

NeutrAvidin Protein, from Invitrogen. Species Reactivity: ; Applications: ELISA, Immunohistochemistry, In Situ Hybridization (ISH), Western Blot.

NeutrAvidin Protein is deglycosylated native avidin from egg whites. Removal of the excess carbohydrate by an exclusive process yields a protein with a more neutral isoelectric point and less nonspecific binding properties. Purified and conjugated forms of NeutrAvidin Protein provide exceptional performance in Western blot, ELISA and IHC applications that require biotin-binding probes. Assay specificity, sensitivity, and signal-to-noise ratios with NeutrAvidin Protein are generally equivalent or better than with the significantly more expensive streptavidin. Avidin is a glycoprotein found in the egg white and tissues of birds, reptiles and amphibia. The biotin-binding protein contains four identical subunits having a combined mass of 67,000-68,000 daltons. Removing the glycosylation from avidin yields NeutrAvidin Protein with a mass of 60,000 daltons. Carbohydrate-based lectin-binding is reduced to undetectable levels, yet biotin-binding affinity is retained. NeutrAvidin Protein offers the advantages of a neutral pI to minimize nonspecific adsorption, along with lysine residues that remain available for derivatization or other customized conjugation. NeutrAvidin Protein yields the lowest nonspecific binding among the known biotin binding proteins. The specific activity for biotin-binding is approximately 14μg/mg of protein, which is near the theoretical maximum activity.

Specifications

Quantity 1000 mg
Content And Storage Upon receipt store product 4°C. Product is shipped at ambient temperature.
Form Solid
Protein Family Avidin
Target Molecule Biotin
Label or Dye Unlabeled
What is the shelf life of NeutrAvidin Protein?

NeutrAvidin Protein is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (4 degrees C). Please see section 8.1 of our Terms & Conditions of Sale for more details.

For Research Use Only. Not for use in diagnostic procedures.