missing translation for 'onlineSavingsMsg'
Learn More

Neutralization solution, Thermo Scientific Chemicals

Numéro de catalogue. AAJ60275AP
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAJ60275AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. AAJ60275AP Fournisseur Thermo Scientific Chemicals Code fournisseur J60275AP
Il en reste null

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Chemical Name or Material Neutralization solution
MDL Number MFCD00283950
Quantity 500 mL
Physical Form Liquid

RUO – Research Use Only