missing translation for 'onlineSavingsMsg'
Learn More

Scigen Neutralex™ Neutra-Pads™ Grossing Pads

Catalog No. 23730570
Change view
Click to view available options
Dimensions (L x W):
8 x 10 in.
11 x 17 in.
Packaging Quantity:
10 Pads/Cs.
100 Pads/Cs.
15 Pads/Cs.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Packaging Quantity
23730570 11 x 17 in. 15 Pads/Cs.
23730568 8 x 10 in. 10 Pads/Cs.
23730567 8 x 10 in. 100 Pads/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23730570 Supplier Scigen Supplier No. 4297
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Neutra-Pads™ instantly neutralizes formalin while rapidly absorbs up to 80 mL of harmful liquid. Blue cut resistance surface with plastic backing keeps your grossing or storage area clean and dry. Certified by the California Environmental Protection Agency.

Neutra-Pads™ were created to absorb and neutralize liquids from large and small specimens during the grossing process. They also are ideal for controlling formalin spills in storage areas and on transportation carts—wherever these liquids must be retained in an enclosed environment.

Designed specifically for use with other Neutralex™ products for maximum effectiveness in treating formalin and aldehyde waste, Neutra-Pads™ have been treated with Neutralex™, which is the only California Environmental Protection Agency-certified solution on the market today.

These pads employ a specially devised backing system that keeps formalin contained and neutralized within the pad itself. The result is a cleaner, drier, and safer environment. Each 8 x 10 in. pad can hold up to 80 mL of liquid.

  • Neutralizes formalin on-contact while absorbing hazardous liquids
  • Highly absorbent polymeric fiber embedded with formalin neutralizer
  • Eliminates hazardous vapors
  • Blue dissection surface consists of a porous highly-resilient polyethylene copolymer cutting area
  • Plastic backing prevents the escape of harmful liquids, keeping you grossing or storage area clean and dry
  • Available in two sizes: 8 x 10 in. or 11 x 17 in.
    • Each 8 x 10 in. pad absorbs up to 80 mL of formalin
    • Each 11 x 17 in. pad absorbs up to 150 mL of formalin
  • Manufactured using ISO 13485 quality system

Specifications

Color Blue
Description Grossing and Storage Pads Instantaneously Neutralize Formalin While Rapidly Absoring Harmful Liquids and Reducing Vapors, Keeps Area Clean and Dry
Capacity (English) 0.02 gal.
Capacity (Metric) 80 mL
Material Pre-treated Formalin Neutralizer, Plastic-backed Absorbent Pad
Type Neutra-Pads™
For Use With (Application) For formalin, grossing and storage safety
Packaging Quantity 15 Pads/Cs.
Dimensions (L x W) 11 x 17 in.