missing translation for 'onlineSavingsMsg'
Learn More

Scigen Neutralex™ Neutra-Pads™ Grossing Pads

Neutra-Pads™ instantly neutralizes formalin while rapidly absorbs up to 80 mL of harmful liquid. Blue cut resistance surface with plastic backing keeps your grossing or storage area clean and dry. Certified by the California Environmental Protection Agency.

Supplier:  Scigen 4295

 View more versions of this product

Catalog No. 23730567

Only null left

Description

Description

Neutra-Pads™ were created to absorb and neutralize liquids from large and small specimens during the grossing process. They also are ideal for controlling formalin spills in storage areas and on transportation carts—wherever these liquids must be retained in an enclosed environment.

Designed specifically for use with other Neutralex™ products for maximum effectiveness in treating formalin and aldehyde waste, Neutra-Pads™ have been treated with Neutralex™, which is the only California Environmental Protection Agency-certified solution on the market today.

These pads employ a specially devised backing system that keeps formalin contained and neutralized within the pad itself. The result is a cleaner, drier, and safer environment. Each 8 x 10 in. pad can hold up to 80 mL of liquid.

  • Neutralizes formalin on-contact while absorbing hazardous liquids
  • Highly absorbent polymeric fiber embedded with formalin neutralizer
  • Eliminates hazardous vapors
  • Blue dissection surface consists of a porous highly-resilient polyethylene copolymer cutting area
  • Plastic backing prevents the escape of harmful liquids, keeping you grossing or storage area clean and dry
  • Available in two sizes: 8 x 10 in. or 11 x 17 in.
    • Each 8 x 10 in. pad absorbs up to 80 mL of formalin
    • Each 11 x 17 in. pad absorbs up to 150 mL of formalin
  • Manufactured using ISO 13485 quality system
Specifications

Chemical Identifiers

Blue
0.02 gal.
Pre-treated Formalin Neutralizer, Plastic-backed Absorbent Pad
For formalin, grossing and storage safety
Grossing and Storage Pads Instantaneously Neutralize Formalin While Rapidly Absoring Harmful Liquids and Reducing Vapors, Keeps Area Clean and Dry
80 mL
Neutra-Pads™

Specifications

Rapid absorption
Grossing and Storage Pads Instantaneously Neutralize Formalin While Rapidly Absoring Harmful Liquids and Reducing Vapors, Keeps Area Clean and Dry
80 mL
Neutra-Pads™
100 Pads/Cs.
Blue
0.02 gal.
Pre-treated Formalin Neutralizer, Plastic-backed Absorbent Pad
For formalin, grossing and storage safety
8 x 10 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions