missing translation for 'onlineSavingsMsg'
Learn More

Neutral Red, Spectrum™ Chemical

$354.14 - $5,588.39

Chemical Identifiers

CAS 553-24-2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18611927
View Documents Promotion Details
N/A 10 g Amber Glass Bottle
Each for $354.14
Only null left
 
18611928
View Documents Promotion Details
N/A 25 g Amber Glass Bottle
Each for $685.86
Only null left
 
18611926
View Documents Promotion Details
N/A 1 kg Amber Glass Bottle
Each for $5,588.39
Only null left
 
Description

Description

Specifications

Chemical Identifiers

553-24-2

Specifications

553-24-2
Reagent
Neutral Red
1
Dark Green
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.