missing translation for 'onlineSavingsMsg'
Learn More

Neutral Red, Spectrum™ Chemical

Catalog No. 18611927
Change view
Click to view available options
Quantity:
10 g
25 g
1 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611927 10 g Amber Glass Bottle
18611928 25 g Amber Glass Bottle
18611926 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18611927 Supplier Spectrum Chemical Mfg Cor Supplier No. NE11310GM

Chemical Identifiers

CAS 553-24-2

Specifications

CAS Max % 1
Grade Reagent
Packaging Amber Glass Bottle
Color Dark Green
Quantity 10 g
Chemical Name or Material Neutral Red

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.