missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neutral Red, Reagent, ACS, Spectrum™ Chemical
$434.68 - $5,234.14
Specifications
| Chemical Name or Material | Neutral Red |
|---|---|
| CAS | 553-24-2 |
| CAS Max % | 1 |
| Grade | ACS |
| Packaging | Amber Glass Bottle |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.