missing translation for 'onlineSavingsMsg'
Learn More

Neutral Red, Reagent, ACS, Spectrum™ Chemical

Catalog No. 18611930
Change view
Click to view available options
Quantity:
10 g
25 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18611930 10 g
18611931 25 g
18611929 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18611930 Supplier Spectrum Chemical Mfg Cor Supplier No. N344510GM

Specifications

Chemical Name or Material Neutral Red
CAS 553-24-2
CAS Max % 1
Grade ACS
Packaging Amber Glass Bottle
Color Dark Green
Quantity 10 g

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.