missing translation for 'onlineSavingsMsg'
Learn More

Neutec Group PS900 Agar/Media Filler

Catalog No. 14910423
Change view
Click to view available options
Capacity:
900 Dishes/Hour (15 mL)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
14910423 900 Dishes/Hour (15 mL)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14910423 Supplier Neutec Group Supplier No. BT900000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PS900 linear filler can fill Up to 900 plates per hour capacity (15mL/Plate)

  • Easy operation software guides smooth transition
  • Dish stacks of required height stack height can be freely selected (1 to 20 dishes/stack)
  • Media smoothing allows you to optimize the filling temperature
  • System utilizes beveled edge/automation plates
  • High sterility at pouring area due to well positioned UV light (with safety switch)
  • Smoother agar plates using dual head pump
  • Repels of agar prevented by an anti drop mechanism
  • Easy operation by friendly touch screen
  • Easy cleaning process due to ergonomic design and easy access
  • Suitable dish diameters: 89 - 96mm
  • Suitable Dish heights: 14 - 20.5mm
  • Delivery rate: 600mL/min.

Specifications

Type Agar/Media Filler
Capacity 900 Dishes/Hour (15 mL)
Temperature (Metric) Operating 5°C to 45°C
Safety Class IP 21
Dimensions (D x W x H) 60 x 18 x 77 cm (23.62 x 70.86 x 30.31 in.)
Weight (English) 176/226 lb.
Weight (Metric) 79.8/102.5 kg
Electrical Requirements 230 VAC - 210mA/360mA, 115 VAC - 310mA/560mA
Frequency 50/60 Hz
Voltage 115 VAC +/-10%, 230 VAC +/-10%