missing translation for 'onlineSavingsMsg'
Learn More

Neurofibromin NF-1 Mouse anti-Rat, Human, Mouse, Clone: NF1-A 376G3, MilliporeSigma™

Mouse Monoclonal Antibody

$284.60 - $731.13

Specifications

Antigen Neurofibromin NF-1
Clone NF1-A 376G3
Concentration 0.5 mg/mL
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABN255725
View Documents Promotion Details
N/A 25 μL
Each for $284.60
Only null left
 
MABN2557100
View Documents Promotion Details
N/A 100 μL
Each for $731.13
Only null left
 
Description

Description

Neurofibromin NF-1 Monoclonal antibody specifically detects Neurofibromin NF-1 in Rat, Human, Mouse samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blotting.
Specifications

Specifications

Neurofibromin NF-1
0.5 mg/mL
Monoclonal
Purified
RUO
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG1 κ
Protein G purified
NF1-A 376G3
Immunocytochemistry, Immunoprecipitation, Western Blot
Unconjugated
Mouse
Rat, Human, Mouse
GST-tagged recombinant fragment corresponding to 369 amino acids from the C-terminal region of human.
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
Videos
SDS
Documents

Documents