missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NeuroD1 Polyclonal Antibody

NeuroD1 Polyclonal Antibody
Supplier: Thermo Fisher Scientific PA5106870
Description
NeuroD1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
| NeuroD1 | |
| Polyclonal | |
| Unconjugated | |
| NEUROD1 | |
| Basic helix-loop-helix factor 1; basic helix-loop-helix transcription factor; Beta2; beta-cell E-box transactivator 2; beta-cell E-box transcriptional activator 2; BHF1; BHF-1; bHLHa3; Class A basic helix-loop-helix protein 3; MODY6; neuro d1; Neurod; Neurod1; neurogenic differentiation 1; neurogenic differentiation factor 1; neurogenic helix-loop-helix protein NEUROD; neuronal differentiation 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 18012, 29458, 4760 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q13562, Q60867, Q64289 | |
| NEUROD1 | |
| A synthesized peptide derived from human NEUROD1(Accession Q13562), corresponding to amino acid residues M1-D31. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |