missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Neural/Glial Antigen 2 (NG2) Monoclonal Antibody (9.2.27), Alexa Fluor™ 488, eBioscience™, Invitrogen™

Catalog No. 50246087
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50246087 25 μg
50246088 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50246087 Supplier Invitrogen™ Supplier No. 53650480
Only null left

Mouse Monoclonal Antibody

Description: This 9.2.27 monoclonal antibody reacts with human neural/glial antigen 2, which is also known as melanoma chondroitin sulfate proteoglycan (MCSP). This antigen is composed of a 250 kDa N-linked glycoprotein and a >450 kDa proteoglycan. NG2 is present on the surface of >90% of maligt melanomas in addition to some non-melanomic tumors. Moreover, NG2 is expressed on glioma cells, as well as on developing and adult oligodendrocyte precursor cells. Studies have demonstrated the involvement of this protein in tumor cell proliferation, adhesion, migration, and invasion. NG2 also functions as a coreceptor for alpha4beta1 integrin. Finally, expression of NG2 can be used as a prognostic marker for disorders such as acral letiginous melanoma and infantile acute myeloid leukemia. The 9.2.27 antibody has been reported to suppress melanoma tumor growth. Applications Reported: This 9.2.27 has been reported for use in flow cytometric analysis, immunohistochemical staining of formalin-fixed paraffin embedded tissue (IHC-P), and immunocytochemical staining of fixed cells (ICC). Applications Tested: This 9.2.27 antibody has been tested by immunocytochemistry on methanol-fixed A375 cells at less than or equal to 20 μg/mL and by flow cytometric analysis on A375 cells at less than or equal to 0.125 μg/test. A test is defined as the amount (ug) of antibody that will stain a cell sample in a final volume of 100 μL.

A human melanoma-associated chondroitin sulfate proteoglycan plays a role in stabilizing cell-substratum interactions during early events of melanoma cell spreading on endothelial basement membranes. CSPG4 represents an integral membrane chondroitin sulfate proteoglycan expressed by human malignant melanoma cells.

Specifications

Antigen Neural/Glial Antigen 2 (NG2)
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Monoclonal
Clone 9.2.27
Concentration 0.5 mg/mL
Conjugate Alexa Fluor 488
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Cspg4
Gene Accession No. Q6UVK1
Gene Alias 4732461B14Rik; AN2; AN2 proteoglycan; AN2/NG2 proteoglycan; chondroitin sulfate proteoglycan 4; chondroitin sulfate proteoglycan 4 (melanoma-associated); chondroitin sulfate proteoglycan NG2; CSPG4; HMW MAA; HMW-MAA; HSN tumor-specific antigen; Kiaa4232; MCSP; MCSPG; MEL CSPG; melanoma chondroitin sulfate proteoglycan; Melanoma-associated chondroitin sulfate proteoglycan; MEL-CSPG; membrane-spanning proteoglycan NG2; MSK16; nerve/glial antigen 2; Ng2; Proteoglycan AN2
Gene Symbols Cspg4
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1464
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only.