missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NeuN Monoclonal Antibody (1B7)
GREENER_CHOICE

Catalog No. PIMA533103
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533103 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533103 Supplier Invitrogen™ Supplier No. MA533103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

NeuN Monoclonal Antibody for ICC/IF, IHC, Western Blot

Fox-3, also known as NEUN, HRNBP3, RBFOX3 (RNA binding protein, fox-1) or Fox-1 homolog C, is a 350 amino acid protein that contains one RRM (RNA recognition motif) domain. Localized to both the nucleus and cytoplasm, Fox-3 is suggested to regulate alternative splicing events. Fox-3 is encoded by a gene located on human chromosome 17, which is comprised over 2.5% of the human genome and encodes over 1,200 genes. Two key tumor suppressor genes are associated with chromosome 17, namely, p53 and BRCA1. Tumor suppressor p53 is necessary for maintenance of cellular genetic integrity by moderating cell fate through DNA repair versus cell death. Malfunction or loss of p53 expression is associated with malignant cell growth and LiFraumeni syndrome. Like p53, BRCA1 is directly involved in DNA repair, though specifically it is recognized as a genetic determinant of early onset breast cancer and predisposition to cancers of the ovary, colon, prostate gland and fallopian tubes.

Specifications

Antigen NeuN
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1B7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 5mM sodium azide
Gene RBFOX3
Gene Accession No. A6NFN3, Q0VD23, Q8BIF2
Gene Alias D11Bwg0517e; FLJ56884; FLJ58356; fox 1 homolog (C. elegans) 3; fox1 homolog C; Fox-1 homolog C; FOX3; FOX-3; FOX3NeuN; hexaribonucleotide binding protein 3; hexaribonucleotide binding protein 3 (HRNBP3); Hexaribonucleotide-binding protein 3; Hrnbp3; NEUN; NeuN antigen; Neuna60; neuronal nuclear antigen A60; neuronal nuclei; Neuronal nuclei antigen; RBFOX3; RFOX3; RGD1560070; RNA binding fox-1 homolog 3; RNA binding protein; RNA binding protein fox-1 homolog 3; RNA binding protein, fox-1 homolog (C. elegans) 3; RNA binding protein, fox-1 homolog 3
Gene Symbols RBFOX3
Host Species Mouse
Immunogen Recombinant protein taken from the N-terminus of human FOX3 expressed in and purified from E. Coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146713, 287847, 511773, 52897
Target Species Human, Mouse, Rat, Bovine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less