missing translation for 'onlineSavingsMsg'
Learn More

Neu5Ac alpha(2-6)Gal beta MP Glycoside 95.0+%, TCI America™

Catalog No. p-7133978
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
Catalog No. Quantity
N079210MG 10 mg
N079250MG 50 mg
2 options

Catalog No. N079210MG

Supplier: TCI America N079210MG

Only null left

Chemical Identifiers

CAS 1984814-41-6
Molecular Formula C24H35NO15
Synonym 4-Methoxyphenyl 6-O-(N-Acetyl-alpha-neuraminosyl)-beta-D-galactopyranoside, 4-Methoxyphenyl O-(5-Acetamido-3,5-dideoxy-D-glycero-alpha-D-galacto-2-nonulopyranosonic Acid)-(2-6)-beta-D-galactopyranoside, Neu5Ac alpha(2-6)Gal-beta-MP

Specifications

Color White
Quantity 10 mg
Formula Weight 577.54
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Neu5Ac alpha(2-6)Gal beta MP Glycoside
TSCA No
Recommended Storage Freezer