CAS RN 1984814-41-6
CAS RN 1984814-41-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Neu5Ac alpha(2-6)Gal beta MP Glycoside 95.0+%, TCI America™
CAS: 1984814-41-6 Molecular Formula: C24H35NO15 Synonym: 4-Methoxyphenyl 6-O-(N-Acetyl-alpha-neuraminosyl)-beta-D-galactopyranoside, 4-Methoxyphenyl O-(5-Acetamido-3,5-dideoxy-D-glycero-alpha-D-galacto-2-nonulopyranosonic Acid)-(2-6)-beta-D-galactopyranoside, Neu5Ac alpha(2-6)Gal-beta-MP