missing translation for 'onlineSavingsMsg'
Learn More

Neu5Ac alpha(2-6)Gal beta MP Glycoside 95.0+%, TCI America™

Catalog No. N079210MG
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N079210MG 10 mg
N079250MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N079210MG Supplier TCI America Supplier No. N079210MG
Only null left

Chemical Identifiers

CAS 1984814-41-6
Molecular Formula C24H35NO15
Synonym 4-Methoxyphenyl 6-O-(N-Acetyl-alpha-neuraminosyl)-beta-D-galactopyranoside, 4-Methoxyphenyl O-(5-Acetamido-3,5-dideoxy-D-glycero-alpha-D-galacto-2-nonulopyranosonic Acid)-(2-6)-beta-D-galactopyranoside, Neu5Ac alpha(2-6)Gal-beta-MP

Specifications

Color White
Quantity 10 mg
Formula Weight 577.54
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Neu5Ac alpha(2-6)Gal beta MP Glycoside
TSCA No
Recommended Storage Freezer