missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neu5Ac alpha(2-3)Gal beta MP Glycoside 95.0+%, TCI America™
$302.25 - $1058.85
Chemical Identifiers
| CAS | 159922-54-0 |
|---|---|
| Molecular Formula | C24H35NO15 |
| Molecular Weight (g/mol) | 577.536 |
| InChI Key | AECZUGNGICKBGW-GGDZMRPFSA-N |
| Synonym | 4-Methoxyphenyl 3-O-(N-Acetyl-alpha-neuraminosyl)-beta-D-galactopyranoside, 4-Methoxyphenyl O-(5-Acetamido-3,5-dideoxy-D-glycero-alpha-D-galacto-2-nonulopyranosonic Acid)-(2-3)-beta-D-galactopyranoside, Neu5Ac alpha(2-3)Gal-beta-MP |
| PubChem CID | 133554172 |
| IUPAC Name | (2R,4S,5S,6S)-5-acetamido-2-[(2R,3R,4R,5R,6R)-3,5-dihydroxy-2-(hydroxymethyl)-6-(4-methoxyphenoxy)oxan-4-yl]oxy-4-hydroxy-6-[(1S,2S)-1,2,3-trihydroxypropyl]oxane-2-carboxylic acid |
| SMILES | CC(=O)NC1C(CC(OC1C(C(CO)O)O)(C(=O)O)OC2C(C(OC(C2O)OC3=CC=C(C=C3)OC)CO)O)O |
Chemical Identifiers
| 159922-54-0 | |
| 577.536 | |
| 4-Methoxyphenyl 3-O-(N-Acetyl-alpha-neuraminosyl)-beta-D-galactopyranoside, 4-Methoxyphenyl O-(5-Acetamido-3,5-dideoxy-D-glycero-alpha-D-galacto-2-nonulopyranosonic Acid)-(2-3)-beta-D-galactopyranoside, Neu5Ac alpha(2-3)Gal-beta-MP | |
| (2R,4S,5S,6S)-5-acetamido-2-[(2R,3R,4R,5R,6R)-3,5-dihydroxy-2-(hydroxymethyl)-6-(4-methoxyphenoxy)oxan-4-yl]oxy-4-hydroxy-6-[(1S,2S)-1,2,3-trihydroxypropyl]oxane-2-carboxylic acid |
| C24H35NO15 | |
| AECZUGNGICKBGW-GGDZMRPFSA-N | |
| 133554172 | |
| CC(=O)NC1C(CC(OC1C(C(CO)O)O)(C(=O)O)OC2C(C(OC(C2O)OC3=CC=C(C=C3)OC)CO)O)O |
Specifications
| 159922-54-0 | |
| C24H35NO15 | |
| 4-Methoxyphenyl 3-O-(N-Acetyl-alpha-neuraminosyl)-beta-D-galactopyranoside, 4-Methoxyphenyl O-(5-Acetamido-3,5-dideoxy-D-glycero-alpha-D-galacto-2-nonulopyranosonic Acid)-(2-3)-beta-D-galactopyranoside, Neu5Ac alpha(2-3)Gal-beta-MP | |
| CC(=O)NC1C(CC(OC1C(C(CO)O)O)(C(=O)O)OC2C(C(OC(C2O)OC3=CC=C(C=C3)OC)CO)O)O | |
| 577.536 | |
| 577.54 | |
| Crystalline Powder |
| White-Yellow | |
| 10 mg | |
| AECZUGNGICKBGW-GGDZMRPFSA-N | |
| (2R,4S,5S,6S)-5-acetamido-2-[(2R,3R,4R,5R,6R)-3,5-dihydroxy-2-(hydroxymethyl)-6-(4-methoxyphenoxy)oxan-4-yl]oxy-4-hydroxy-6-[(1S,2S)-1,2,3-trihydroxypropyl]oxane-2-carboxylic acid | |
| 133554172 | |
| ≥95.0% (HPLC) | |
| Neu5Ac alpha(2-3)Gal beta MP Glycoside |
Safety and Handling
TSCA : No
Recommended Storage : Freezer