CAS RN 159922-54-0
CAS RN 159922-54-0
IUPAC Name:
2-{[3,5-dihydroxy-2-(hydroxymethyl)-6-(4-methoxyphenoxy)oxan-4-yl]oxy}-5-acetamido-4-hydroxy-6-(1,2,3-trihydroxypropyl)oxane-2-carboxylic acid
Synonyms:
2-{[3,5-dihydroxy-2-(hydroxymethyl)-6-(4-methoxyphenoxy)oxan-4-yl]oxy}-5-acetamido-4-hydroxy-6-(1,2,3-trihydroxypropyl)oxane-2-carboxylic acid
Molecular Weight (g/mol):
577.54
Molecular Formula:
C24H35NO15
InChi Key:
AECZUGNGICKBGW-UHFFFAOYNA-N
SMILES:
COC1=CC=C(OC2OC(CO)C(O)C(OC3(CC(O)C(NC(C)=O)C(O3)C(O)C(O)CO)C(O)=O)C2O)C=C1
1
–
1
of
1
results
1
Neu5Ac alpha(2-3)Gal beta MP Glycoside 95.0+%, TCI America™
CAS: 159922-54-0 Molecular Formula: C24H35NO15 Molecular Weight (g/mol): 577.536 InChI Key: AECZUGNGICKBGW-GGDZMRPFSA-N Synonym: 4-Methoxyphenyl 3-O-(N-Acetyl-alpha-neuraminosyl)-beta-D-galactopyranoside, 4-Methoxyphenyl O-(5-Acetamido-3,5-dideoxy-D-glycero-alpha-D-galacto-2-nonulopyranosonic Acid)-(2-3)-beta-D-galactopyranoside, Neu5Ac alpha(2-3)Gal-beta-MP PubChem CID: 133554172 IUPAC Name: (2R,4S,5S,6S)-5-acetamido-2-[(2R,3R,4R,5R,6R)-3,5-dihydroxy-2-(hydroxymethyl)-6-(4-methoxyphenoxy)oxan-4-yl]oxy-4-hydroxy-6-[(1S,2S)-1,2,3-trihydroxypropyl]oxane-2-carboxylic acid SMILES: CC(=O)NC1C(CC(OC1C(C(CO)O)O)(C(=O)O)OC2C(C(OC(C2O)OC3=CC=C(C=C3)OC)CO)O)O