missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nessler Reagent, for Ammonia Nitrogen Determination, Ricca Chemical
Ammonia Nitrogen Determination
$159.47 - $1,123.43
Specifications
| Color | Clear light yellow |
|---|---|
| Format | Liquid |
| Sterility | Non-sterile |
| Type | Nessler Reagent |
| Catalog Number | Mfr. No. | Volume (Metric) | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume (Metric) | Packaging | Price | Quantity & Availability | ||||
|
R52500004B
|
N/A | 4 L | Black Poly Bottle |
Each for $1,123.43
|
|
||||
|
525032
|
N/A | 1 L | Amber Poly Bottle |
Each for $482.07
|
|
||||
|
52504
|
N/A | 120 mL | Amber Poly Bottle |
Each for $159.47
|
|
||||
|
525016
|
N/A | 500 mL | Amber Poly Bottle |
Each for $306.00
|
|
||||
Description
- For Ammonia Nitrogen Determination
Specifications
| Clear light yellow | |
| Liquid | |
| Non-sterile | |
| Nessler Reagent |