missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nessler Reagent, for Ammonia Nitrogen Determination, Ricca Chemical
Ammonia Nitrogen Determination
Supplier: Ricca Chemical Company 525016
Description
- For Ammonia Nitrogen Determination
Chemical Identifiers
| Clear light yellow | |
| Liquid | |
| Non-sterile | |
| Nessler Reagent |
Specifications
| Clear light yellow | |
| Liquid | |
| 500 mL | |
| Nessler Reagent |
| 500 mL | |
| Amber Poly Bottle | |
| Non-sterile | |
| 16.9 oz. |