missing translation for 'onlineSavingsMsg'
Learn More

Nervonic acid, ≥99% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014371
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014371 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014371 Supplier Merck Emd Millipore Supplier No. 87117100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Nervonic acids are the products of desaturation and elongation processes of various fatty acids such as palmitic acid, stearic acid, oleic acid, etc.

Specifications

CAS 506-37-6
Flash Point 110°C - closed cup
Shipping Condition Ambient
Physical Form Neat
Linear Formula CH3(CH2)7 CH=CH(CH2)13 COOH (cis)
MDL Number MFCD00010507
Melting Point 42°C to 43°C
Synonym cis-15-Tetracosenoic acid; Selacholeic acid
Type Analytical Standard
Quantity 100 mg
Formula Weight 366.62
Percent Purity ≥99% (GC)
Grade Analytical Standard
Show More Show Less