missing translation for 'onlineSavingsMsg'
Learn More

Nerolidol, cis + trans, 97+%

Catalog No. AAA1881214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1881214 25 g
AAA1881222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1881214 Supplier Thermo Scientific Chemicals Supplier No. A1881214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7212-44-4 | C15H26O | 222.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7212-44-4
Molecular Formula C15H26O
Molecular Weight (g/mol) 222.37
MDL Number MFCD00008911,MFCD00008911,MFCD00008911,MFCD00085350
InChI Key FQTLCLSUCSAZDY-SDNWHVSQNA-N
Synonym 3r,6e-nerolidol, unii-uoc0644v25, 3r-6e-nerolidol, nerolidol, 3r,6e-3,7,11-trimethyldodeca-1,6,10-trien-3-ol, e-nerolidol, 1,6,10-dodecatrien-3-ol, 3,7,11-trimethyl-, 3r,6e, nerolidol, 6e--, --nerolidol, ?-nerolidol
PubChem CID 11241545
ChEBI CHEBI:59959
SMILES CC(C)=CCC\C(C)=C\CCC(C)(O)C=C

Specifications

Density 0.876
Boiling Point 263°C to 266°C
Flash Point 96°C (204°F)
Refractive Index 1.479
Quantity 25 g
Beilstein 1724135
Merck Index 14,6477
IUPAC Name (3R,6E)-3,7,11-trimethyldodeca-1,6,10-trien-3-ol
Formula Weight 222.37
Percent Purity ≥97%
Chemical Name or Material Nerolidol, cis + trans
Show More Show Less
Hazard Category H319
Precautionary Statement P264b-P280i-P305+P351+P338
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 230-597-5
RTECSNumber JR4977000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only