missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nerol 98.0+%, TCI America™
$72.15 - $271.51
Chemical Identifiers
| CAS | 106-25-2 |
|---|---|
| Molecular Formula | C10H18O |
| Molecular Weight (g/mol) | 154.25 |
| MDL Number | MFCD00063204 |
| InChI Key | GLZPCOQZEFWAFX-YFHOEESVSA-N |
| Synonym | nerol, cis-geraniol, neryl alcohol, z-3,7-dimethylocta-2,6-dien-1-ol, z-geraniol, 2z-3,7-dimethylocta-2,6-dien-1-ol, 2,6-octadien-1-ol, 3,7-dimethyl-, 2z, nerol natural, cis-3,7-dimethyl-2,6-octadien-1-ol, z-3,7-dimethyl-2,6-octadien-1-ol |
| PubChem CID | 643820 |
| ChEBI | CHEBI:29452 |
| IUPAC Name | (2Z)-3,7-dimethylocta-2,6-dien-1-ol |
| SMILES | CC(C)=CCC\C(C)=C/CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
N007725ML
|
N/A | 25 mL |
Each for $72.15
|
|
|||||
|
N0077500ML
|
N/A | 500 mL |
Each for $271.51
|
|
|||||
Chemical Identifiers
| C10H18O | |
| MFCD00063204 | |
| nerol, cis-geraniol, neryl alcohol, z-3,7-dimethylocta-2,6-dien-1-ol, z-geraniol, 2z-3,7-dimethylocta-2,6-dien-1-ol, 2,6-octadien-1-ol, 3,7-dimethyl-, 2z, nerol natural, cis-3,7-dimethyl-2,6-octadien-1-ol, z-3,7-dimethyl-2,6-octadien-1-ol | |
| CHEBI:29452 | |
| CC(C)=CCC\C(C)=C/CO |
Specifications
| 106-25-2 | |
| 226°C | |
| MFCD00063204 | |
| GLZPCOQZEFWAFX-YFHOEESVSA-N | |
| (2Z)-3,7-dimethylocta-2,6-dien-1-ol | |
| 643820 | |
| 154.25 | |
| Liquid |
| Yellow | |
| C10H18O | |
| nerol, cis-geraniol, neryl alcohol, z-3,7-dimethylocta-2,6-dien-1-ol, z-geraniol, 2z-3,7-dimethylocta-2,6-dien-1-ol, 2,6-octadien-1-ol, 3,7-dimethyl-, 2z, nerol natural, cis-3,7-dimethyl-2,6-octadien-1-ol, z-3,7-dimethyl-2,6-octadien-1-ol | |
| CC(C)=CCC\C(C)=C/CO | |
| 154.25 | |
| CHEBI:29452 | |
| ≥98.0% (GC) | |
| Nerol |
Safety and Handling
EINECSNumber : (2)-0258
RTECSNumber : RG5840000
TSCA : Yes