missing translation for 'onlineSavingsMsg'
Learn More

Nerol 98.0+%, TCI America™

Catalog No. N007725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N007725ML 25 mL
N0077500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N007725ML Supplier TCI America Supplier No. N007725ML

Chemical Identifiers

CAS 106-25-2
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00063204
InChI Key GLZPCOQZEFWAFX-YFHOEESVSA-N
Synonym nerol, cis-geraniol, neryl alcohol, z-3,7-dimethylocta-2,6-dien-1-ol, z-geraniol, 2z-3,7-dimethylocta-2,6-dien-1-ol, 2,6-octadien-1-ol, 3,7-dimethyl-, 2z, nerol natural, cis-3,7-dimethyl-2,6-octadien-1-ol, z-3,7-dimethyl-2,6-octadien-1-ol
PubChem CID 643820
ChEBI CHEBI:29452
IUPAC Name (2Z)-3,7-dimethylocta-2,6-dien-1-ol
SMILES CC(C)=CCC\C(C)=C/CO

Specifications

Color Yellow
Boiling Point 226°C
Quantity 25 mL
Formula Weight 154.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Nerol
EINECSNumber (2)-0258
RTECSNumber RG5840000
TSCA Yes