missing translation for 'onlineSavingsMsg'
Learn More

Nerol, 97%, Thermo Scientific Chemicals

$88.63

Chemical Identifiers

CAS 106-25-2
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00063204
InChI Key GLZPCOQZEFWAFX-YFHOEESVSA-N
Synonym nerol, cis-geraniol, neryl alcohol, z-3,7-dimethylocta-2,6-dien-1-ol, z-geraniol, 2z-3,7-dimethylocta-2,6-dien-1-ol, 2,6-octadien-1-ol, 3,7-dimethyl-, 2z, nerol natural, cis-3,7-dimethyl-2,6-octadien-1-ol, z-3,7-dimethyl-2,6-octadien-1-ol
PubChem CID 643820
ChEBI CHEBI:29452
IUPAC Name (2Z)-3,7-dimethylocta-2,6-dien-1-ol
SMILES CC(C)=CCC\C(C)=C/CO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAA19808AE
View Documents Promotion Details
N/A 100 mL
Each for $88.63
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

106-25-2
154.25
GLZPCOQZEFWAFX-YFHOEESVSA-N
643820
(2Z)-3,7-dimethylocta-2,6-dien-1-ol
C10H18O
MFCD00063204
nerol, cis-geraniol, neryl alcohol, z-3,7-dimethylocta-2,6-dien-1-ol, z-geraniol, 2z-3,7-dimethylocta-2,6-dien-1-ol, 2,6-octadien-1-ol, 3,7-dimethyl-, 2z, nerol natural, cis-3,7-dimethyl-2,6-octadien-1-ol, z-3,7-dimethyl-2,6-octadien-1-ol
CHEBI:29452
CC(C)=CCC\C(C)=C/CO

Specifications

106-25-2
103°C to 105°C (9 mmHg)
C10H18O
MFCD00063204
14,6475
GLZPCOQZEFWAFX-YFHOEESVSA-N
(2Z)-3,7-dimethylocta-2,6-dien-1-ol
643820
154.25
Nerol
0.87
77°C (171°F)
1.475
1722455
nerol, cis-geraniol, neryl alcohol, z-3,7-dimethylocta-2,6-dien-1-ol, z-geraniol, 2z-3,7-dimethylocta-2,6-dien-1-ol, 2,6-octadien-1-ol, 3,7-dimethyl-, 2z, nerol natural, cis-3,7-dimethyl-2,6-octadien-1-ol, z-3,7-dimethyl-2,6-octadien-1-ol
CC(C)=CCC\C(C)=C/CO
154.25
CHEBI:29452
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 203-378-7

RTECSNumber : RG5840000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only