missing translation for 'onlineSavingsMsg'
Learn More

Nerol, 97%, Thermo Scientific Chemicals

Catalog No. AAA19808AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA19808AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA19808AE Supplier Thermo Scientific Chemicals Supplier No. A19808AE
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 106-25-2 | Molecular Formula: C10H18O | Molecular Weight: 154.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 106-25-2
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00063204
InChI Key GLZPCOQZEFWAFX-YFHOEESVSA-N
Synonym nerol, cis-geraniol, neryl alcohol, z-3,7-dimethylocta-2,6-dien-1-ol, z-geraniol, 2z-3,7-dimethylocta-2,6-dien-1-ol, 2,6-octadien-1-ol, 3,7-dimethyl-, 2z, nerol natural, cis-3,7-dimethyl-2,6-octadien-1-ol, z-3,7-dimethyl-2,6-octadien-1-ol
PubChem CID 643820
ChEBI CHEBI:29452
IUPAC Name (2Z)-3,7-dimethylocta-2,6-dien-1-ol
SMILES CC(C)=CCC\C(C)=C/CO

Specifications

Density 0.87
Boiling Point 103°C to 105°C (9 mmHg)
Flash Point 77°C (171°F)
Refractive Index 1.475
Quantity 100 mL
Beilstein 1722455
Merck Index 14,6475
Formula Weight 154.25
Percent Purity 97%
Chemical Name or Material Nerol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 203-378-7
RTECSNumber RG5840000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only