missing translation for 'onlineSavingsMsg'
Learn More

Nerol, ≥97% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012391
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012391 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012391 Supplier Merck Emd Millipore Supplier No. 509491ML

missing translation for 'validMainframeMsg'

Nerol belongs to the class of monoterpene glycosides.

Specifications

CAS 106-25-2
Density 0.876 g/mL (at 25°C)
Flash Point 107°C
Physical Form Neat
Linear Formula (CH3)2 C=CHCH2 CH2 C(CH3)=CHCH2 OH
MDL Number MFCD00063204
Boiling Point 103°C to 105°C (9 mmHg)
Synonym cis-3,7-Dimethyl-2,6-octadien-1-ol; Nerol
Type Analytical Standard
Quantity 1 mL
Formula Weight 154.25
Percent Purity ≥97% (GC)
Grade Analytical Standard
Show More Show Less