missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ NERL™ Trutol™ Glucose Tolerance Test Beverages
Screen for diabetes mellitus, gestational diabetes and hypoglycemia with these glucose tolerance test beverages.
$142.45
Specifications
| Certifications/Compliance | Kosher |
|---|---|
| Content And Storage | 15°C to 30°C |
| Detectable Analytes | None |
| Type | Reagent |
| Packaging Type | 24 Bottles/Cs. |
| Catalog Number | Mfr. No. | Flavor | Dose | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Flavor | Dose | Price | Quantity & Availability | ||||
|
TGP401223PA
|
Thermo Scientific™
401223PA |
Orange | 75g |
Case of 24 Each for $142.45
|
|
||||
Description
Screen for diabetes mellitus, gestational diabetes and hypoglycemia with Thermo Scientific™ NERL™ Trutol™ Glucose Tolerance Test Beverages. Beverages are noncarbonated, caffeine-free and available in 50, 75 and 100g concentrations.
- Noncarbonated beverages provide easier consumption, a reduction of digestive discomfort and a more palatable taste
- Graduated bottles assure proper dose every time
- Kosher-certified
Ordering Alerts:
- Plastic bottles packaged in cases of 24 bottles; four six-packs per case
Specifications
| Kosher | |
| None | |
| 24 Bottles/Cs. |
| 15°C to 30°C | |
| Reagent | |
| 10 oz. |
Safety and Handling
ShelfLife : 24 Months