missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NERL™ Trutol™ Glucose Tolerance Test Beverages

Catalog No. TGP401223PA
Change view
Click to view available options
Flavor:
Orange
Dose:
75g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flavor Dose
TGP401223PA Orange 75g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TGP401223PA Supplier Thermo Scientific™ Supplier No. 401223PA
Only null left

Screen for diabetes mellitus, gestational diabetes and hypoglycemia with these glucose tolerance test beverages.

Screen for diabetes mellitus, gestational diabetes and hypoglycemia with Thermo Scientific™ NERL™ Trutol™ Glucose Tolerance Test Beverages. Beverages are noncarbonated, caffeine-free and available in 50, 75 and 100g concentrations.

  • Noncarbonated beverages provide easier consumption, a reduction of digestive discomfort and a more palatable taste
  • Graduated bottles assure proper dose every time
  • Kosher-certified

Ordering Alerts:

  • Plastic bottles packaged in cases of 24 bottles; four six-packs per case

Specifications

Certifications/Compliance Kosher
Content And Storage 15°C to 30°C
Detectable Analytes None
Type Reagent
Dose 75g
Flavor Orange
Packaging Type 24 Bottles/Cs.
Quantity 10 oz.