missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nepsilon-Trifluoroacetyl-L-lysine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 254°C |
| Color | White |
| Quantity | 5 g |
| Synonym | h-lys tfa-oh, n-6-trifluoroacetyl-l-lysine, n6-trifluoroacetyl-l-lysine, n6-trifluoroaccety-l-lysine, epsilon-tfa-lysine, nepsilon-trifluoroacetyl-l-lysine, l-lysine, n6-trifluoroacetyl, n 6-trifluoroacetyl-l-lysine, n-, a-trifluoroacetyl-l-lysine |
| Formula Weight | 242.20 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Nepsilon-Trifluoroacetyl-L-lysine |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |