missing translation for 'onlineSavingsMsg'
Learn More

Nepsilon-(tert-Butoxycarbonyl)-Nalpha-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-lysine 98.0+%, TCI America™

Supplier:  TCI America B307225G

 View more versions of this product

Catalog No. B307225G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Nepsilon-(tert-Butoxycarbonyl)-Nalpha-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-lysine
124°C
C26H32N2O6
25 g
UMRUUWFGLGNQLI-ANBDAQEENA-N
(2S)-6-{[(tert-butoxy)carbonyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
2724628
≥98.0% (HPLC,T)
71989-26-9
White-Yellow
MFCD000371
fmoc-lys boc-oh, fmoc-lys boc, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-tert-butoxycarbonyl amino hexanoic acid, n-alpha-fmoc-nepsilon-boc-l-lysine, nepsilon-boc-nalpha-fmoc-l-lysine, n-alpha-fmoc-nepsilon-boc-l-lysine, n-fmoc-n'-boc-l-lysine, nepsilon-tert-butoxycarbonyl-nalpha-9h-fluoren-9-ylmethoxy carbonyl-l-lysine, n, a-fmoc-n, n, a-fmoc-n
CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
468.55
468.55
Crystalline Powder

Chemical Identifiers

71989-26-9
468.55
UMRUUWFGLGNQLI-ANBDAQEENA-N
2724628
CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
C26H32N2O6
MFCD000371
fmoc-lys boc-oh, fmoc-lys boc, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-tert-butoxycarbonyl amino hexanoic acid, n-alpha-fmoc-nepsilon-boc-l-lysine, nepsilon-boc-nalpha-fmoc-l-lysine, n-alpha-fmoc-nepsilon-boc-l-lysine, n-fmoc-n'-boc-l-lysine, nepsilon-tert-butoxycarbonyl-nalpha-9h-fluoren-9-ylmethoxy carbonyl-l-lysine, n, a-fmoc-n, n, a-fmoc-n
(2S)-6-{[(tert-butoxy)carbonyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions